Learn More
Thermo Scientific™ Clear Strong Heat Sealing Film
Product Code. 11969355
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 100
Product Code. 11969355
Brand: Thermo Scientific™ AB0685
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Transparent—suitable for genotyping applications
  • Clear polyester polypropylene laminate heat seal
  • Strong and unpeelable
  • Also suitable for storage and disposal of hazardous material
  • Good solvent resistance; recommended for use with DMSO
  • Seal integrity range: -80°C to +120°C
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Sterility Non-sterile