missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Luminaris HiGreen qPCR Master Mix, fluorescein
Thermo Scientific Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
266.00 € - 3815.00 €
Specifications
Detection Method | SYBR |
---|---|
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Reaction Speed | Standard |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
13565260
|
Thermo Scientific™
K0984 |
5000 Reactions |
3815.00 €
5000 reactions |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13545260
|
Thermo Scientific™
K0981 |
250 Reactions |
266.00 €
250 reactions |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13535260
|
Thermo Scientific™
K0982 |
500 Reactions |
510.00 €
500 reactions |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13555260
|
Thermo Scientific™
K0983 |
1250 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
SYBR | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression | |
2 X |
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
qPCR Master Mix | |
2X | |
DNA, dsDNA |