Learn More
Thermo Scientific™ GLP Compliance Services for Plasma Blast Freezer
Descrizione
IQ
- Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
- Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ System Component Information Detailed configuration information for each system component have been recorded.
- OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
- Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Door Ajar, Power Failure, Temperature Alerts
Specifica
Specifica
| For Use With (Equipment) | Thermo Scientific Blast Freezer including Thermo Scientific XBF |
| Description | Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. |
| Product Type | GLP Compliance Services for Plasma Blast Freezer |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | Includes travel to major metropolitan areas. Does not include Installation. Instrument must be installed powered up and set temperature point for 24 hours before on-site service |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?