missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ Open-top Tank Liner System, 200L
Shop All Thermo Scientific ProductsDescription
Tank Liner Key Features:
- Fits into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Conical Bottom Drums
- Gusset design allows for a close fit to the tank
- Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific HyPerforma BPCs
- Supplied gamma irradiated
- Customization options are available by contacting one of our Thermo Scientific Representatives today
Applications:
Hydration of powdered media and buffers; Pooling of nonsterile solutions and fluids
Order Info
Supplied double polybagged and irradiated with Certificate of Analysis.
Specifications
Specifications
| Capacity (Metric) | 200 L |
| Material | Plastic |
| Diameter (English) | 23 in. |
| Product Type | Tank Liner |
| Quantity | Each |
| For Use With (Equipment) | HyPerforma Conical Bottom Drum |
| Sterilization Method | Gamma Irradiated |
| Tubing Line 1 | 182.9 cm (72 in) |
| Tubing Line 2 | None |
| Product Line | Thermo Scientific |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?