Learn More
Thermo Scientific™ MIC 2000 Microtiter microplate
GREENER_CHOICE
Product Code. 10558105
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
Product Code. 10558105
Brand: Thermo Scientific™ 120151
Only null left
Add to basket
Add to basket
For support of microbiolgical applications
These products are suitable for applications including bacterial growth, bacterial identification, minimum inhibitory concentration (MIC), hybridization, hybridoma plating and cloning.
  • Microtiter microplate format
  • Polystyrene
  • U-bottom
Order Info
Bag of 20; Box of 100
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.28 mL
Well Shape Round Bottom (U-well)