Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Service for Thermo Scientific™ Barnstead Smart2Pure Pro
Product Code. 17478733
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Barnstead Smart2Pure Pro
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17478733 Thermo Scientific Barnstead Smart2Pure Pro Each
1 options
Product Code. 17478733 Supplier Unity Lab Services™ Supplier No. IOQPCKE50133921
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Water Purification Validation Service for Thermo Scientific Barnstead Smart2Pure Pro
IQ
  • Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Flow Adjustment, Water Pressure, Resistivity, Power Failure
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Barnstead Smart2Pure Pro
Description Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing.
Service Offering Industry-Exclusive, No-Charge Requalification Guarantee. Unity Lab Services is the only OEM service provider to offer a guarantee and promise to requalify instruments and equipment at no charge if a key component fails while under the original factory warranty or a qualifying instrument service plan.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service