missing translation for 'onlineSavingsMsg'
Learn More

Thiamethoxam 1000 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30316132
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316132 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316132 Supplier Dr. Ehrenstorfer Supplier No. DREA17453000TO1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiamethoxam
CAS 153719-23-4
Analyte or Component Names Thiamethoxam
Shipping Condition Room Temperature
Molecular Formula C8 H10 Cl N5 O3 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 4H-1,3,5-Oxadiazin-4-imine, 3-[(2-chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro-, 3-[(2-Chloro-5-thiazolyl)methyl]tetrahydro-5-methyl-N-nitro-4H-1,3,5-oxadiazin-4-imine, A 97565N, A 9765N, Actara, Actara 25WG, Actara 2GR, Adage, Adage 5FS, CGA 293343, Centric, Cruiser, Cruiser 350FS, Cruiser 5FS, Cruiser 600FS, Cruiser A 9765, Diacloden, FS 350, FS 600, FS 600 (insecticide), Flagship, Meridian, Optigard, Platinum (insecticide), Platinum 240SC, Thiamethoxam, WS 70, WS 70 (insecticide), Xamox
InChI Formula InChI=1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3
SMILES CN1COCN(Cc2cnc(Cl)s2)C1=N[N+](=O)[O-]
IUPAC Name N-[3-[(2-chloro-1,3-thiazol-5-yl)methyl]-5-methyl-1,3,5-oxadiazinan-4-ylidene]nitramide
Solvent or Matrix Toluene
Molecular Weight (g/mol) 291.71
Quantity 1 mL
Formula Weight 291.0193 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.