missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, 99.5%, MP Biomedicals™

Product Code. 11475251 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475251 25 g 25g
11465251 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475251 Supplier MP Biomedicals Supplier No. 0215684625
  • ONLINE EXCLUSIVE PRICE
    650.00 €
    616.35 € / 25g
    Save 33.65 €
    5% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 15318-45-3
Molecular Formula C12H15Cl2NO5S
Molecular Weight (g/mol) 356.214
InChI Key OTVAEFIXJLOWRX-NXEZZACHSA-N
Synonym D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide
PubChem CID 27200
ChEBI CHEBI:32215
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

Type Thiamphenicol
Physical Form Free-flowing Powder
Color White
Quantity 25 g
Assay Percent Range 0.995
Formula Weight 356.2
Grade Not Available
Chemical Name or Material Thiamphenicol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.