missing translation for 'onlineSavingsMsg'
Learn More

Thimerosal, Fisher BioReagents

Product Code. 10657155
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass
Unit Size:
100g
10g
25g
500g
Product Code. Quantity Packaging unitSize
10657155 100 g Amber Glass 100g
10356793 10 g Amber Glass 10g
10234823 25 g Amber Glass 25g
10506204 500 g Amber Glass 500g
4 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Product Code. 10657155

Brand: Fisher BioReagents BP2542100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
  • Thimerosal is a mercury-containing compound commonly used as an antibacterial and antifungal agent
  • Also used as a bactericide in pharmaceuticals and research
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym Sodium ethylmercurithiosalicylate
PubChem CID 16684434
ChEBI CHEBI:9546
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Type Thimerosal
Physical Form Solid
pH 6.7 to 8.2
Melting Point 232°C
Loss on Drying 0.5% max.
Packaging Amber Glass
Color Cream
Quantity 100 g
Assay Percent Range ≥97 %
Merck Index 15, 9470
Formula Weight 404.8
Percent Purity 97% min.
Chemical Name or Material Thimerosal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thimerosal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.