missing translation for 'onlineSavingsMsg'
Learn More

Thimerosal

Product Code. 15424319
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424319 25 g 25g
15414319 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424319 Supplier Thermo Scientific Alfa Aesar Supplier No. J61799.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Thimerosal, CAS # 54-64-8, is a sulfhydryl (thiol) oxidizing reagent with antibacterial and antifungal activities that is used to protect components from chemical change or microbial action. | CAS: 54-64-8 | C9H9HgNaO2S | 404.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Thimerosal is an organic compound and a derivative of thiosalicyclic acid. It shows antibacterial and antifungal activities and has the ability to protect against chemical change or microbial action

• Thimerosal can inhibit the sulfhydryl-containing active site of some enzymes and bind to sulfhydryl compounds, such as glutathione, cysteine, and thiol groups of proteins

• It can activate the InsP3 calcium channel on the endoplasmic reticular membrane, triggering the release of calcium from intracellular stores, leading to a calcium-influx of extracellular calcium. Thus, thimerosal may stimulate or inhibit cellular functions dependent on calcium signaling

Application

• Thimerosal can increase histamine release and cell-mediated immunity

• It has the ability to inhibit various sulfhydryl-dependent enzymes and proteins, including PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na+/K+-dependent ATPases

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 234°C to 237°C
Color White
Flash Point 250°C (482°F)
Quantity 25 g
UN Number UN2025
Beilstein 8169555
Merck Index 14,9317
Solubility Information Soluble in water or ethanol
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
Formula Weight 404.81
Physical Form Powder
Chemical Name or Material Thimerosal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thimerosal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium o-(ethylmercurithio)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.