missing translation for 'onlineSavingsMsg'
Learn More

Thio-TEPA, MedChemExpress

Product Code. 16477376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477376 Supplier Medchem Express Supplier No. HY17574
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Thio-TEPA is a DNA alkylating agent, with antitumor activity.

Thio-TEPA is a DNA alkylating agent, with antitumor activity.

Specifications

Chemical Name or Material Thio-TEPA
Physical Form Solid
CAS 52-24-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H12N3PS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (264.24 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES S=P(N1CC1)(N2CC2)N3CC3
Molecular Weight (g/mol) 189.22
Formula Weight 189.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.