missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbanilide, 98%

Product Code. 13428358 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
100g
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13428358 2.5 kg Plastic Drum 2.5kg
10666722 100 g Plastic bottle 100g
10051640 500 g Plastic bottle 500g
10656722 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13428358 Supplier Thermo Scientific Acros Supplier No. 138700025
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-08-9 | C13H12N2S | 228.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-08-9
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00004921
InChI Key FCSHMCFRCYZTRQ-UHFFFAOYSA-N
Synonym thiocarbanilide, diphenylthiourea, 1,3-diphenyl-2-thiourea, n,n'-diphenylthiourea, sulfocarbanilide, sym-diphenylthiourea, thiourea, n,n'-diphenyl, stabilisator c, rhenocure ca, vulkacit ca
PubChem CID 700999
IUPAC Name 1,3-diphenylthiourea
SMILES S=C(NC1=CC=CC=C1)NC1=CC=CC=C1

Specifications

Melting Point 151.0°C to 154.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic Drum
Linear Formula C6H5NHCSNHC6H5
Quantity 2.5 kg
Beilstein 12,394
Merck Index 15,3373
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 228.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Thiocarbanilide, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thiocarbanilide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.