missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic anhydride, 98%

Product Code. 11485383
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11485383 5 g 5g
11495383 25 g 25g
11405393 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11485383 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A13390.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 3261-87-8 | C4H4O3S | 132.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3261-87-8
Molecular Formula C4H4O3S
Molecular Weight (g/mol) 132.133
MDL Number MFCD00051689
InChI Key RIIUAPMWDSRBSH-UHFFFAOYSA-N
Synonym thiodiglycolic anhydride, 1-oxa-4-thia-cyclohexan-2,6-dione
PubChem CID 137871
IUPAC Name 1,4-oxathiane-2,6-dione
SMILES C1C(=O)OC(=O)CS1

Specifications

Melting Point 95°C to 100°C
Formula Weight 132.13
Percent Purity 98%
Quantity 5 g
Sensitivity Moisture sensitive
Chemical Name or Material Thiodiglycolic anhydride
compliance-icons
Product Identifier
  • Thiodiglycolic anhydride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Thiodiglycolate anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.