missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Thiol Peroxidase Protein

Product Code. p-200038887 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18449590

Brand: Novus Biologicals™ NBP1788050.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of E.coli Thiol Peroxidase The Recombinant E. coli Thiol Peroxidase Protein is derived from E. coli. The Recombinant E. coli Thiol Peroxidase Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_415840
Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2 mM DTT, 0.1 M NaCl.
Molecular Weight (g/mol) 19.9 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant E. coli TPX protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TPX, 1-168 aa, E. coli, His tag, E. coli (NP_415840).
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Thiol Peroxidase
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.