missing translation for 'onlineSavingsMsg'
Learn More

Thionyl chloride, 99.7%, Thermo Scientific Chemicals

Product Code. 10138472 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
Product Code. Quantity unitSize
10138472 2.5 L 2.5 L
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10138472

Brand: Thermo Scientific Acros 382660025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7719-09-7
Molecular Formula Cl2OS
Molecular Weight (g/mol) 118.96
InChI Key FYSNRJHAOHDILO-UHFFFAOYSA-N
IUPAC Name sulfurooyl dichloride
SMILES ClS(Cl)=O

Specifications

Melting Point -105.0°C
Density 1.6400 g/mL
Boiling Point 76.0°C
Assay Percent Range 99.7%
Packaging Glass bottle
Linear Formula SOCl2
Refractive Index 1.5190 to 1.5210
Quantity 2.5 L
Merck Index 15, 9501
Specific Gravity 1.64
Solubility Information Solubility aq. soln.: reacts. Other solubilities: miscible with benzene, chloroform and carbon, tetrachloride
Viscosity 0.6 mPa.s (20°C)
Formula Weight 118.97
Chemical Name or Material Thionyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.