missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Thiophosphate ester Antibody (SR4347), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27854-50ul
This item is not returnable.
View return policy
Description
Thiophosphate ester Monoclonal antibody specifically detects Thiophosphate ester in Nonspecies specific samples. It is validated for Immunoprecipitation
Specifications
| Thiophosphate ester | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| A synthesized peptide derived from human Thiophosphate ester | |
| 50 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation | |
| SR4347 | |
| Immunoprecipitation 1:1000 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction