missing translation for 'onlineSavingsMsg'
Learn More

Thioxanthene, 98%

Product Code. 10762054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762054 1 g Glass bottle 1g
10024673 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10762054 Supplier Thermo Scientific Acros Supplier No. 391670010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 261-31-4 | C13H10S | 198.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261-31-4
Molecular Formula C13H10S
Molecular Weight (g/mol) 198.28
MDL Number MFCD00046936
InChI Key PQJUJGAVDBINPI-UHFFFAOYSA-N
Synonym thioxanthene, 10h-dibenzo b,e thiin, thiaxanthen, thioxanthen, dibenzothiapyran, 9h-thioxanthen, 10h-dibenzo b,e thiopyran, chembl79451
PubChem CID 67495
ChEBI CHEBI:51055
IUPAC Name 9H-thioxanthene
SMILES C1C2=CC=CC=C2SC2=CC=CC=C12

Specifications

Melting Point 128°C to 131°C
Color White
Boiling Point 340°C (730.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 9522
Solubility Information Solubility in water: .. Other solubilities: freely soluble in chloroform,moderately soluble in alcohol,ether
Formula Weight 198.29
Percent Purity 98%
Physical Form Crystals or Needles
Chemical Name or Material Thioxanthene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.