accessibility menu, dialog, popup

UserName

CAS RN 261-31-4

S

CAS RN 261-31-4

IUPAC Name: 9H-thioxanthene
Synonyms: thioxanthene thioxanthine
Molecular Weight (g/mol): 198.28
Molecular Formula: C13H10S
InChi Key: PQJUJGAVDBINPI-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2SC2=CC=CC=C12
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results