missing translation for 'onlineSavingsMsg'
Learn More

Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™
BIOPHARMA

Product Code. 12916193 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12916193 100 mL 100mL
12906193 50 mL 50mL
12926193 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12916193 Supplier Fisher Chemical Supplier No. MS 007 100
  • ONLINE EXCLUSIVE PRICE
    365.00 €
    329.68 € / 100mL
    Save 35.32 €
    9% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Thirty one elements solution - Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb Se Sm Sr, Tl, Tm, U, V, Zn for ICP/MS 10mg/L HNO3 matrix

Specifications

Chemical Name or Material Thirty-One Elements
Packaging Bottle
Analyte or Component Names Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P, Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn
Physical Form Liquid
Concentration 10 mg/L
Type ICP/MS Calibration Multi-element Standard Solution
Element Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb, Se, Sm, Sr, TI, Tm, U, V, Zn
Solvent or Matrix 2% HNO3
Quantity 100 mL
Product Identifier
  • Multi-element Standard Solution for ICP. 31 Components in HNO3 2% (MS 007)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.