missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOP1 Mouse, anti-Human, Clone: 1B11A11, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | THOP1 |
|---|---|
| Clone | 1B11A11 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18035615
|
Proteintech
68587-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039885
|
Proteintech
68587-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The THOP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human THOP1. This antibody recognizes human, mouse , rat antigen. The THOP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| THOP1 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, Thimet oligopeptidase, thimet oligopeptidase 1, THOP1, TOP | |
| Primary |
| 1B11A11 | |
| Western Blot, ELISA | |
| Mouse | |
| 7064 | |
| IgG2a | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title