missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thrombomodulin Monoclonal Antibody (4D8.2C6)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA562071
This item is not returnable.
View return policy
Description
Thrombomodulin Monoclonal Antibody for Western Blot, IHC (P), Flow
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
Specifications
| Thrombomodulin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P15306 | |
| THBD | |
| Two synthetic peptides made to internal portions of the mouse Thrombomodulin protein (between residues 25-150 and 400-550). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 4D8.2C6 | |
| Unconjugated | |
| THBD | |
| AHUS6; AI385582; BDCA3; BDCA-3; CD141; CD141 antigen; DC141; fetomodulin; sCD141; snoRNA MBII-339; soluble CD141; THBD; THPH12; THRM; thrombomdulin; thrombomodulin; thrombomodulin precursor; TM | |
| Rat | |
| Protein G | |
| RUO | |
| 21824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction