missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thrombospondin 1 Recombinant Mouse Monoclonal Antibody (20B2)

Description
Thrombospondin 1 Recombinant Monoclonal Antibody for Flow, ELISA
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Specifications
Specifications
| Antigen | Thrombospondin 1 |
| Applications | ELISA, Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 20B2 |
| Concentration | 1.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | THBS1 |
| Gene Accession No. | P07996 |
| Gene Alias | glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?