missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRSP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98508-100ul
This item is not returnable.
View return policy
Description
THRSP Polyclonal antibody specifically detects THRSP in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| THRSP | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| lipogenic protein 1, Lpgp, LPGP1, S14MGC21659, Spot 14 protein, SPOT14SPOT14 homolog, thyroid hormone responsive, thyroid hormone responsive (SPOT14 homolog, rat), thyroid hormone responsive SPOT14, thyroid hormone responsive SPOT14 (rat) homolog, thyroid hormone-inducible hepatic protein | |
| Produced in rabbits immunized with E. coli-derived Human THRSP fragment. | |
| 100 μg | |
| metabolism | |
| 7069 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction