missing translation for 'onlineSavingsMsg'
Learn More

Thulium Standard: Tm @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17908679
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908679 100 mL 100mL
17909149 250 mL 250mL
17988669 50 mL 50mL
17939149 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17908679 Supplier VHG Labs Supplier No. VHGPTMN100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Thulium Standard: Tm @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Thulium
CAS 7440-30-4
Methods ICP/ICP-MS
Analyte or Component Names Thulium
Shipping Condition Room Temperature
Molecular Formula Tm
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Tm
InChI Formula InChI=1S/Tm
SMILES [Tm]
IUPAC Name thulium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 168.934
Quantity 100 mL
Formula Weight 168.934 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.