missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thymidine Phosphorylase Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148475

Product Code. 30533705

  • 479.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thymidine Phosphorylase
Polyclonal
Unconjugated
TYMP
2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP
Goat
Antigen affinity chromatography
RUO
1890
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry (Paraffin), Neutralization, Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
P19971
TYMP
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PD-ECGF/Thymidine Phosphorylase, Ala11-Gln482.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.