missing translation for 'onlineSavingsMsg'
Learn More

Thymidine Phosphorylase / PD-ECGF Mouse, Clone: SPM322, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1890-MSM1X-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929917

  • 462.00 € / 100µg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Thymidine Phosphorylase / PD-ECGF
Cytoplasmic and Nuclear
SPM322
Unconjugated
NP_001107227
Mouse
55kDa
Protein A/G Purified
RUO
1890
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
TYMP
Recombinant full-length human Thymidine Phosphorylase (TP / PD-ECGF) protein
HUVEC cells. Breast, Bladder, Lung or Kaposi tumors
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers