Learn More
Thyroid Cancer Marker, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Thyroid Cancer Marker, Mouse Monoclonal Antibody.
146.00 € - 647.70 €
Specifications
| Antigen | Thyroid Cancer Marker |
|---|---|
| Clone | 373E1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12650457
|
Epredia
MS-1914-S0 |
100 μL | Concentrated | ||||||
|
12670457
|
Epredia
MS-1914-S1 |
500 μL | Concentrated | ||||||
|
12640457
|
Epredia
MS-1914-S |
1 mL | Concentrated | ||||||
|
12660457
|
Epredia
MS-1914-R7 |
7 mL | Prediluted | ||||||
Description
Papillary carcinoma is by far the most frequent malignant tumor of the thyroid with a threefold prevelance in females. Studies have shown that 100% of malignant thyroid lesions were found to contain the keratan sulfate (KS) bearing molecules, while these were virtually absent from benign tissues and other thyroid tumors, with the exception of 21% of follicular carcinoma cases analyzed. Clone 373E1 is specific for the keratan sulfate bearing molecular complex.
Host Species: Mouse
Clone: 373E1
Species Reactivity: Human. Others not tested.
Positive Control: Papillary Thyroid CA
Cellular Localization: Membrane, cytoplasmic and luminal
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| Thyroid Cancer Marker | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Papillary Thyroid CA |
| 373E1 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.