Learn More
Thyroid Cancer Marker, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Thyroid Cancer Marker, Mouse Monoclonal Antibody.
Brand: Epredia MS-1914-R7
Description
Papillary carcinoma is by far the most frequent malignant tumor of the thyroid with a threefold prevelance in females. Studies have shown that 100% of malignant thyroid lesions were found to contain the keratan sulfate (KS) bearing molecules, while these were virtually absent from benign tissues and other thyroid tumors, with the exception of 21% of follicular carcinoma cases analyzed. Clone 373E1 is specific for the keratan sulfate bearing molecular complex.
Host Species: Mouse
Clone: 373E1
Species Reactivity: Human. Others not tested.
Positive Control: Papillary Thyroid CA
Cellular Localization: Membrane, cytoplasmic and luminal
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| Thyroid Cancer Marker | |
| Monoclonal | |
| Unconjugated | |
| Papillary Thyroid CA | |
| RUO | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| 373E1 | |
| Mouse | |
| 7 mL | |
| Cancer and Tumor Biology | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.