missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid Peroxidase Antibody (TPO/7088R), Novus Biologicals™
Rabbit Monoclonal Antibody
298.00 € - 538.00 €
Specifications
| Antigen | Thyroid Peroxidase |
|---|---|
| Clone | TPO/7088R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680244
|
Novus Biologicals
NBP3-20268-100ug |
100 μg |
538.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657304
|
Novus Biologicals
NBP3-20268-20ug |
20 μg |
298.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Thyroid Peroxidase | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 10mM PBS with 0.05% BSA | |
| 7173 | |
| IgG κ | |
| Protein A or G purified |
| TPO/7088R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Recombinant fragment of human Thyroid Peroxidase (around aa 600-900) (Exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title