Learn More
Thyroid Peroxidase Antibody (TPO28), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-50811AF647
Product Code. 18203185
  • 452.00 € / 0.25mL
Description
Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thyroid Peroxidase
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
TPO
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human thyroid peroxidase.
0.25 mL
Lipid and Metabolism
7173
Store at 4C in the dark.
IgG1
Western Blot, ELISA
TPO28
Western Blot, ELISA, Sandwich ELISA
EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
Mouse
Protein A purified
RUO
Primary
Human
Purified