missing translation for 'onlineSavingsMsg'
Learn More

Thyroid Peroxidase Mouse anti-Human, HRP, Antibody Pair, Novus Biologicals™

Product Code. 18375693 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18375693 15 Plates Pack of 15
18330942 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18375693 Supplier Novus Biologicals Supplier No. NBP30670815PLATES
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody pair for Solid phase sandwich ELISA

Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for ELISA/Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroid Peroxidase
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Gene Alias EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
Host Species Mouse
Quantity 15 Plates
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Type Antibody Pairs
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.