Learn More
Invitrogen™ TIA-1 Recombinant Rabbit Monoclonal Antibody (24GB2175)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557777
 View more versions of this product
Product Code. 30276226
  • 198.00 € / 20µL
Description
TIA-1 Recombinant Monoclonal Antibody for Western Blot, Flow
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIA-1
Recombinant Monoclonal
Unconjugated
TIA1
2310050N03Rik; AI256674; cytotoxic granule-associated RNA binding protein 1; cytotoxic granule-associated RNA-binding protein 1; mTIA-1; Nucleolysin TIA-1; nucleolysin TIA-1 isoform p40; p40 TIA 1; p40-TIA-1; p40-TIA-1 (containing p15-TIA-1); RNA-binding protein TIA-1; T-cell-restricted intracellular antigen-1; Tia; TIA 1; TIA1; TIA-1; TIA1 cytotoxic granule-associated RNA binding protein; TIA1 protein; TIAL1; TIAR; WDM
Rabbit
Affinity chromatography
RUO
21841, 312510, 7072
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2175
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P31483, P52912
TIA1
A synthesized peptide derived from human TIA1 (aa 324-386).
20 μL
Primary
Human, Mouse, Rat