missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIA1 Antibody (rTIA1/1353), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20584-100ug
This item is not returnable.
View return policy
Description
TIA1 Monoclonal antibody specifically detects TIA1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TIA1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein | |
| Recombinant full-length human TIA1 protein | |
| 100 μg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 7072 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rTIA1/1353 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction