missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIA1 Mouse, Clone: TIA1/1313, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | TIA1 |
---|---|
Clone | TIA1/1313 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990968
|
EnquireBio
7072-MSM1-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980968
|
EnquireBio
7072-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
TIA1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7072 | |
Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
40kDa |
TIA1/1313 | |
Western Blot | |
Unconjugated | |
RUO | |
XP_016860286 | |
TIA1 | |
IgG2b | |
Protein A/G Purified | |
Cytoplasmic | |
HepG2, PC3 and HeLa cells. Tonsil. |