missing translation for 'onlineSavingsMsg'
Learn More

TIC10 isomer, MedChemExpress

Product Code. 16486836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486836

Brand: Medchem Express HY15615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TIC10 isomer is the isomer of TIC10. TIC10 isomer does not possess the reported biological activity of inducing TRIAL expression.

TIC10 isomer is the isomer of TIC10. TIC10 isomer does not possess the reported biological activity of inducing TRIAL expression.

Specifications

Chemical Name or Material TIC10 isomer
CAS 41276-02-2
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H26N4O
Synonym ONC201 isomer
Solubility Information DMSO : 5 mg/mL (12.94 mM; Need ultrasonic)
SMILES O=C(C(C1)=C(N2CC3=C(C)C=CC=C3)CCN1CC4=CC=CC=C4)N5C2=NCC5
Molecular Weight (g/mol) 386.49
Formula Weight 386.49
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.