missing translation for 'onlineSavingsMsg'
Learn More

Ticagrelor TCG Sulfonate, TRC

Product Code. 30449184
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449184 10 mg 10mg
30450081 25 mg 25mg
30449295 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449184 Supplier Toronto Research Chemicals Supplier No. TRCT43772510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ticagrelor TCG Sulfonate
Analyte or Component Names Ticagrelor TCG Sulfonate
Shipping Condition Room Temperature
Molecular Formula C23 H28 F2 N6 O7 S2
Synonym 2-(((1S,2S,3S,4R)-4-(7-(((1R,2S)-2-(3,4-difluorophenyl)cyclopropyl)amino)-5-(propylthio)-3H-[1,2,3]triazolo[4,5-d]pyrimidin-3-yl)-2,3-dihydroxycyclopentyl)oxy)ethyl hydrogen sulfate
InChI Formula InChI=1S/C23H28F2N6O7S2/c1-2-7-39-23-27-21(26-15-9-12(15)11-3-4-13(24)14(25)8-11)18-22(28-23)31(30-29-18)16-10-17(20(33)19(16)32)37-5-6-38-40(34,35)36/h3-4,8,12,15-17,19-20,32-33H,2,5-7,9-10H2,1H3,(H,26,27,28)(H,34,35,36)/t12-,15+,16+,17-,19-,20+/m0/s1
Solution Type Neat
SMILES CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](OCCOS(=O)(=O)O)[C@@H](O)[C@H]5O)c4n1
IUPAC Name 2-[(1S,2S,3S,4R)-4-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyl-triazolo[4,5-d]pyrimidin-3-yl]-2,3-dihydroxy-cyclopentoxy]ethyl hydrogen sulfate
Molecular Weight (g/mol) 602.63
Quantity 10 mg
Formula Weight 602.1429
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.