missing translation for 'onlineSavingsMsg'
Learn More

Tideglusib, MedChemExpress

Product Code. 16446576
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446576 100 mg 100mg
16426228 10 mg 10mg
16412357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16446576 Supplier Medchem Express Supplier No. HY14872
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tideglusib (NP031112) is an irreversible GSK-3 inhibitor with IC50s of 5 nM and 60 nM for GSK-3βWT (1 h preincubation) and GSK-3βC199A (1 h preincubation), respectively.

Tideglusib (NP031112) is an irreversible GSK-3 inhibitor with IC50s of 5 nM and 60 nM for GSK-3βWT (1 h preincubation) and GSK-3βC199A (1 h preincubation), respectively.

Specifications

Chemical Name or Material Tideglusib
Physical Form Solid
CAS 865854-05-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H14N2O2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym NP031112
Solubility Information DMSO : 33.33 mg/mL (99.67 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N(SC1=O)C2=C3C=CC=CC3=CC=C2)N1CC4=CC=CC=C4
Molecular Weight (g/mol) 334.39
Formula Weight 334.39
Percent Purity 99.52%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.