missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie-1 Antibody (8D12D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
187.00 € - 557.00 €
Specifications
| Antigen | Tie-1 |
|---|---|
| Clone | 8D12D2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161099
|
Novus Biologicals
NBP2-37267 |
0.1 mL |
557.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620666
|
Novus Biologicals
NBP2-37267-0.025ml |
0.025 mL |
187.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tie-1 Monoclonal specifically detects Tie-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Tie-1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
| TIE1 | |
| IgG1 | |
| Affinity Purified |
| 8D12D2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| 7075 | |
| Purified recombinant fragment of human TIE1 (AA: 385-607) expressed in E. Coli. | |
| Primary | |
| 125 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title