missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGAR/C12orf5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 361.00 €
Specifications
| Antigen | TIGAR/C12orf5 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18149001
|
Novus Biologicals
NBP2-24754SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705103
|
Novus Biologicals
NBP2-24754 |
361.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TIGAR/C12orf5 | |
| Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q9NQ88 | |
| 57103 | |
| A portion of amino acids 140-190 of human TIGAR was used as the immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein | |
| TIGAR | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human and 93% homologous in chimp. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title