missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT, Armenian Hamster anti-Mouse, Clone: 4D4, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-81116-0.2MG
This item is not returnable.
View return policy
Description
TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry, Block/Neutralize.
Specifications
| TIGIT | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, Block/Neutralize | |
| T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| 201633 | |
| Mouse | |
| Purified |
| Flow Cytometry, Blocking Assay | |
| 4D4 | |
| Unconjugated | |
| PBS | |
| TIGIT | |
| Armenian hamsters were immunized with recombinant mouse TIGIT tetramers by a combination of s.c. and food pad immunization and booster injections. Draining lymph nodes were fused with Sp2/0-Ag14 cells, selected in HAT (hypoxanthine/aminopterin/thymidine) medium and supernatants were screened for specificity by ELISA and flow cytometry using TIGIT-transfectants. | |
| 0.2 mg | |
| Primary | |
| 4D4 specifically recognises TIGIT. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction