Learn More
Invitrogen™ TIGIT Monoclonal Antibody (GIGD7), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
180.00 € - 407.00 €
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | GIGD7 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16370584
|
Invitrogen™
11-9501-80 |
25 μg |
180.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16380584
|
Invitrogen™
11-9501-82 |
100 μg |
407.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GIGD7 monoclonal antibody reacts with mouse T cell immunoreceptor with Ig and ITIM domains (TIGIT, VSTM3, WUCAM), a 26 kDa protein that is a member of the poliovirus receptor (PVR) family. In mice, the expression of TIGIT has been reported in follicular T helper cells, while in humans, the expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells. TIGIT is not expressed on B cells or naive CD4+ T cells. TIGIT is upregulated on CD4+ T cells following activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK cells and T cells with antigen presenting cells, fibroblasts and endothelial cells that express PVR and PVR-like proteins. This GIGD7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
| TIGIT | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| TIGIT | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| GIGD7 | |
| Flow Cytometry | |
| FITC | |
| Rat | |
| Mouse | |
| P86176 | |
| 100043314 | |
| mTIGIT-Fc treated with 4% PFA. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TIGIT |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.