missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-2 Antibody (RMT2-14) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
250.00 € - 705.00 €
Specifications
| Antigen | TIM-2 |
|---|---|
| Clone | RMT2-14 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672179
|
Novus Biologicals
NBP3-09036-0.025mg |
0.025 mg |
250.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618276
|
Novus Biologicals
NBP3-09036-0.2mg |
0.2 mg |
705.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-2 Monoclonal antibody specifically detects TIM-2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional AssaySpecifications
| TIM-2 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| T Cell immunoglobulin and mucin domain protein 2, Tim2 | |
| IgG2a λ | |
| Protein A purified |
| RMT2-14 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Functional Assay | |
| Unconjugated | |
| Rat | |
| Cellular Signaling, Immunology | |
| PBS | |
| This antibody was raised by immunising Sprague Dawley rats with TIM-2-Ig consisting of the extracellular domain (aa 1-230 of mouse TIM-2) and the Fc portion of mouse IgG2a, emulsified in complete Freund's adjuvant and then fusing lymph node cells from the immunized rats with P3U1 myeloma cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title