missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, PE, Clone: 7D3, BD Pharmingen™
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
| Antigen | TIM-3 (CD366) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 7D3 |
| Conjugate | PE |
| Description | Vol. Per test: 5μL |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 |
| Host Species | Mouse |
| Immunogen | Human TIM-3 |
| Show More |
For Reseach Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?