missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM50 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94349-0.02ml
This item is not returnable.
View return policy
Description
TIMM50 Polyclonal antibody specifically detects TIMM50 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TIMM50 | |
| Polyclonal | |
| Western Blot 1:1000-1:4000 | |
| homolog of yeast Tim50, MGC102733, mitochondrial import inner membrane translocase subunit TIM50, TIM50, TIM50L, Tim50-like protein, translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae), translocase of inner mitochondrial membrane 50 homolog (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-456 of human TIMM50 (NP_001001563.1). GNNPVDENGAKIPDEFDNDPILVQQLRRTYKYFKDYRQMIIEPTSPCLLPDPLQEPYYQPPYTLVLELTGVLLHPEWSLATGWRFKKRPGIETLFQQLAPLYEIVIFTSETGMTAFPLIDSVDPHGFISYRLFRDATRYMDGHHVKDISCLNRDPARVVVVDCKKEAFRLQPYNGVALRPWDGNSDDRVLLDLSAFLKTIALNGVEDVRTVLEHYALEDDPLAAFKQRQSRLEQEEQQRLAELSKSNKQNLFLGSLTSRLWPRSKQP | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 92609 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction