missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM50 Antibody (SR1606), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22096-50ul
This item is not returnable.
View return policy
Description
TIMM50 Monoclonal antibody specifically detects TIMM50 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| TIMM50 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1606 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| homolog of yeast Tim50, MGC102733, mitochondrial import inner membrane translocase subunit TIM50, TIM50, TIM50L, Tim50-like protein, translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae), translocase of inner mitochondrial membrane 50 homolog (yeast) | |
| A synthesized peptide derived from human TIMM50 (Uniprot #: Q3ZCQ8) | |
| 50 μg | |
| Endocrinology, Signal Transduction | |
| 92609 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction