Learn More
Invitrogen™ TIMM50 Recombinant Rabbit Monoclonal Antibody (24GB2670)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557780
 View more versions of this product
Product Code. 30275039
  • 564.00 € / 100µL
Description
TIMM50 Recombinant Monoclonal Antibody for Western Blot, Flow
TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TIMM50
Recombinant Monoclonal
Unconjugated
TIMM50
2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357
Rabbit
Affinity chromatography
RUO
66525, 92609
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2670
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q3ZCQ8, Q9D880
TIMM50
A synthesized peptide derived from human TIMM50 (aa 25-77).
100 μL
Primary
Human, Mouse