missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20645
This item is not returnable.
View return policy
Description
TIMP-4 Polyclonal specifically detects TIMP-4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| TIMP-4 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
| Rabbit | |
| 26 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Hypoxia | |
| 7079 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.46mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q99727 | |
| TIMP4 | |
| Recombinant protein encompassing a sequence within the center region of human TIMP4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on sequence identity: Mouse (91%), Monkey (99%), Rabbit (95%), Rat (92%), Bovine (91%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction