missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIN2 Antibody (OTI2C12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45024-100ul
This item is not returnable.
View return policy
Description
TIN2 Monoclonal antibody specifically detects TIN2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| TIN2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 26277 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI2C12 | |
| Western Blot 1:500-2000, Immunohistochemistry 1:2000 | |
| (TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 266-347 of human TINF2 (NP_036593) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction