missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tinidazole

Product Code. 16594384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594384 5 g 5g
16504394 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594384 Supplier Thermo Scientific Chemicals Supplier No. 463510050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19387-91-8
Molecular Formula C8H13N3O4S
Molecular Weight (g/mol) 247.27
MDL Number MFCD00057217
InChI Key HJLSLZFTEKNLFI-UHFFFAOYSA-N
IUPAC Name 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
SMILES CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O

Specifications

Melting Point 124.0°C to 128.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, slightly soluble in methanol, very slightly soluble on ethanol
Formula Weight 247.27
Chemical Name or Material Tinidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.