accessibility menu, dialog, popup

UserName

CH3SOONCH3NNOO

CAS RN 19387-91-8

IUPAC Name: 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
Synonyms: tinidazole tindamax fasigyn timidazole
Molecular Weight (g/mol): 247.27
Molecular Formula: C8H13N3O4S
InChi Key: HJLSLZFTEKNLFI-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results